Learn More
(16a)-21-(Acetyloxy)-16-methyl-pregna-1,4,9(11)-triene-3,20-dione, TRC
Product Code. 30310913
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30310913 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30310913 Supplier Toronto Research Chemicals Supplier No. TRCA18742525MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (16alpha)-21-(Acetyloxy)-16-methyl-pregna-1,4,9(11)-triene-3,20-dione
CAS 4258-83-7
Molecular Formula C24H30O4
Quantity 25 mg
Synonym 21-Acetoxy-16alpha-methylpregna-1,4,9-(11)-triene-3,20-dione
InChI Formula InChI=1S/C24H30O4/c1-14-11-20-18-6-5-16-12-17(26)7-9-23(16,3)19(18)8-10-24(20,4)22(14)21(27)13-28-15(2)25/h7-9,12,14,18,20,22H,5-6,10-11,13H2,1-4H3/t14-,18-,20+,22?,23+,24+/m1/s1
SMILES C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)C3=CC[C@]2(C)C1C(=O)COC(=O)C
IUPAC Name [2-oxo-2-[(8S,10S,13S,14S,16R)-10,13,16-trimethyl-3-oxo-6,7,8,12,14,15,16,17-octahydrocyclopenta[a]phenanthren-17-yl]ethyl] acetate
Molecular Weight (g/mol) 382.49
Formula Weight 382.2144