Learn More
Greiner Bio-One 1536-well Low Base Polystyrene Microplates
Product Code. 13037983
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color
13037983 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13037983 Supplier Greiner Bio-One Supplier No. 783856
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
For optical applications in high-content screening. Greiner Bio-One 1536 Well Low Base Polystyrene Microplates enables interference-free measurements and also microscopy over whole surface of bottom of microplate.
  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic
  • Improved geometry
  • Short distance of 0.3 mm between external well bottom and microplate skirt bottom
  • Facilitates changing objectives during microscopy
  • TRUSTED_SUSTAINABILITY
Specifications
Format Standard
Description Model: Sensoplate Plus
Array 32 x 48
Color Black
Material Polystyrene
No. of Wells 1536
Sterility Non-sterile
Well Shape Flat Bottom (F-well)