missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC6A12 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67700-1-IG-150UL
This item is not returnable.
View return policy
Description
The SLC6A12 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SLC6A12. This antibody recognizes human, mouse, rat antigen. The SLC6A12 antibody has been validated for the following applications: WB, IF/ICC, ELISA analysis.Specifications
| SLC6A12 | |
| Monoclonal | |
| 1700 μg/mL | |
| BGT-1; BGT1; FLJ38727 | |
| Recombinant Protein | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| 70 kDa | |
| Liquid |
| Western Blot, Immunocytochemistry/Immunofluorescence, ELISA | |
| 2C1G4 | |
| Unconjugated | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 6539 | |
| Store at -20°C. Stable for one year after shipment. | |
| Monoclonal Antibody | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction