missing translation for 'onlineSavingsMsg'
Learn More

15-Azido-4,7,10,13-tetraoxapentadecanoic acid

Product Code. p-7042861
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15476159 25 mg 25mg
15486159 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15476159 Supplier Thermo Scientific Alfa Aesar Supplier No. J64030.MB
Add to basket
This item is not returnable. View return policy

CAS: 1257063-35-6 | C11H21N3O6 | 291.30 g/mol

Azido-PEG4-Acid is amine- or phosphine-reactive building block used to derivative primary amines for ligation to alkyne-tagged molecules.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Azido-PEG4-Acid is amine- or phosphine-reactive building block used to derivative primary amines for ligation to alkyne-tagged molecules.

Solubility
Soluble in water.

Notes
Protect from heat. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1257063-35-6
Molecular Formula C11H21N3O6
Molecular Weight (g/mol) 291.30
MDL Number MFCD12406155
InChI Key BODPHGOBXPGJKO-UHFFFAOYSA-N
Synonym azido-peg4-acid, 15-azido-4,7,10,13-tetraoxapentadecanoic acid, n3-peg 4-cooh, 1-azido-3,6,9,12-tetraoxapentadecan-15-oic acid, azido-dpeg r 4-acid, 3-2-2-2-2-azidoethoxy ethoxy ethoxy ethoxy propanoic acid
PubChem CID 22731907

Specifications

Color Colorless to Yellow
Linear Formula N3CH2CH2(OCH2CH2)4CO2H
Quantity 25 mg
Solubility Information Soluble in water.
Formula Weight 291.3
Physical Form Liquid
Chemical Name or Material 15-Azido-4,7,10,13-tetraoxapentadecanoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.