Learn More
Welch Ilmvac™ 14 Laborzentrifugen Oil
Shop All Welch Ilmvac Products
Click to view available options
Volume (Metric):
1 L
2 L
5 L
10 L
Unit Size:
1 L
10 L
2 L
5 L
Specifications
Specifications
| Product Type | Oil |
| For Use With (Equipment) | Rotary Vane Pumps |
| Volume (Metric) | 10 L |