Learn More
12-Heptacosanone, TRC
Specifications
Specifications
| Chemical Name or Material | 12-Heptacosanone |
| CAS | 31534-85-7 |
| Molecular Formula | C27H54O |
| Quantity | 100 mg |
| Synonym | 12-Heptacosanone |
| InChI Formula | InChI=1S/C27H54O/c1-3-5-7-9-11-13-14-15-16-18-20-22-24-26-27(28)25-23-21-19-17-12-10-8-6-4-2/h3-26H2,1-2H3 |
| SMILES | CCCCCCCCCCCCCCCC(=O)CCCCCCCCCCC |
| IUPAC Name | heptacosan-12-one |
| Molecular Weight (g/mol) | 394.72 |
| Formula Weight | 394.4175 |