Learn More
12-Heptacosanone, TRC
Product Code. 30399327
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30399327 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30399327 Supplier Toronto Research Chemicals Supplier No. TRCH789345250MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 12-Heptacosanone
CAS 31534-85-7
Molecular Formula C27H54O
Quantity 250 mg
Synonym 12-Heptacosanone
InChI Formula InChI=1S/C27H54O/c1-3-5-7-9-11-13-14-15-16-18-20-22-24-26-27(28)25-23-21-19-17-12-10-8-6-4-2/h3-26H2,1-2H3
SMILES CCCCCCCCCCCCCCCC(=O)CCCCCCCCCCC
IUPAC Name heptacosan-12-one
Molecular Weight (g/mol) 394.72
Formula Weight 394.4175