Learn More
11b,21-dihydroxypregna-1,4,16-triene-3,20-dione, TRC
Specifications
Specifications
| Chemical Name or Material | 11beta,21-dihydroxypregna-1,4,16-triene-3,20-dione |
| CAS | 3103-17-1 |
| Molecular Formula | C21H26O4 |
| Quantity | 50 mg |
| Synonym | (11beta)-11,21-Dihydroxypregna-1,4,16-triene-3,20-dione, (8S,9S,10R,11S,13S,14S)-11-Hydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-6,7,8,9,10,11,12,13,14,15-decahydro-3H-cyclopenta[a]phenanthren-3-one, Prednisolone Impurity |
| InChI Formula | InChI=1S/C21H26O4/c1-20-8-7-13(23)9-12(20)3-4-14-15-5-6-16(18(25)11-22)21(15,2)10-17(24)19(14)20/h6-9,14-15,17,19,22,24H,3-5,10-11H2,1-2H3/t14-,15-,17-,19+,20-,21-/m0/s1 |
| SMILES | C[C@]12C[C@H](O)[C@H]3[C@@H](CCC4=CC(=O)C=C[C@]34C)[C@@H]1CC=C2C(=O)CO |
| Molecular Weight (g/mol) | 342.43 |
| Formula Weight | 342.1831 |