Learn More
11-Cyclopropyl-5,11-dihydro-4-(hydroxymethyl)-6H-dipyrido[3,2-b:2',3'-e][1,4]diazepin-6-one-d2, TRC
Specifications
Specifications
| Chemical Name or Material | 11-Cyclopropyl-5,11-dihydro-4-(hydroxymethyl)-6H-dipyrido[3,2-b:2',3'-e][1,4]diazepin-6-one-d2 |
| CAS | 2733558-78-4 |
| Molecular Formula | C15H12D2N4O2 |
| Quantity | 1 mg |
| Synonym | 11-Cyclopropyl-4-(hydroxymethyl)-5H-dipyrido[3,2-b:2',3'-e][1,4]diazepin-6(11H)-one-d2 |
| InChI Formula | InChI=1S/C15H14N4O2/c20-8-9-5-7-17-14-12(9)18-15(21)11-2-1-6-16-13(11)19(14)10-3-4-10/h1-2,5-7,10,20H,3-4,8H2,(H,18,21)/i8D2 |
| SMILES | O=C1NC(C(C([2H])([2H])O)=CC=N2)=C2N(C3CC3)C4=C1C=CC=N4 |
| IUPAC Name | 11-cyclopropyl-4-(hydroxymethyl-d2)-5,11-dihydro-6H-dipyrido[3,2-b:2',3'-e][1,4]diazepin-6-one |
| Molecular Weight (g/mol) | 284.31 |
| Formula Weight | 284.12 |