Learn More
11-Bromoundecanoic Acid, TRC
Specifications
Spezifikation
| Chemical Name or Material | 11-Bromoundecanoic Acid |
| CAS | 2834-05-1 |
| Molecular Formula | C11H21BrO2 |
| Quantity | 2.5 g |
| Synonym | NSC 14781, ω-Bromoundecanoic Acid |
| InChI Formula | InChI=1S/C11H21BrO2/c12-10-8-6-4-2-1-3-5-7-9-11(13)14/h1-10H2,(H,13,14) |
| SMILES | OC(=O)CCCCCCCCCCBr |
| IUPAC Name | 11-bromoundecanoic acid |
| Molecular Weight (g/mol) | 265.19 |
| Formula Weight | 264.0725 |