missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ 10nm Colloidal Gold Nanoparticles 1OD

Product Code. 18040744 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 mL
100 mL
Unit Size:
100mL
20mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18040744 100 mL 100mL
18031086 20 mL 20mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18040744 Supplier Novus Biologicals™ Supplier No. 30010100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

10nm colloidal gold nanoparticles ultra high quality, with narrow size distribution and uniform shape

  • 10nm colloidal gold nanoparticles are ultra high quality, with narrow size distribution and uniform shape, supplied in range of sizes and concentrations for maximum flexibility
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 4°C. Do not freeze.
Quantity 100 mL
Kit Contents One 20mL bottle at 1 OD
Product Type 10nm Colloidal Gold Nanoparticles 1OD Antibody Labeling Kit

For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.