missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ PTC 209

Product Code. 18745268
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18745268

Brand: Tocris Bioscience™ 5191/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Bmi-1 inhibitor; antitumor

PTC 209 is a Bmi-1 inhibitor (IC50 ∽ 0.5 μM); irreversibly impairs colorectal cancer-initiating cell (CIC) growth. Reduces tumor growth in CIC xenograft assays and results in reduced potential of colorectal cancer cells to initiate tumors in vivo.
TRUSTED_SUSTAINABILITY

Specifications

Purity >98%
Quantity 10 mg
Product Type PTC 209
Molecular Weight (g/mol) 495.19
Inhibitors PTC 209

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.