missing translation for 'onlineSavingsMsg'
Learn More

NVP-HSP990, MedChemExpress

Product Code. 16426358
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16426358 10 mg 10mg
16462467 50 mg 50mg
16418947 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16426358 Supplier Medchem Express Supplier No. HY15190

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NVP-HSP990 is a potent and selective Hsp90 inhibitor, with IC50 values of 0.6, 0.8, and 8.5 nM for Hsp90α, Hsp90β, and Grp94, respectively.

NVP-HSP990 is a potent and selective Hsp90 inhibitor, with IC50 values of 0.6, 0.8, and 8.5 nM for Hsp90α, Hsp90β, and Grp94, respectively.

Specifications

Chemical Name or Material NVP-HSP990
CAS 934343-74-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C20H18FN5O2
Quantity 10 mg
Synonym HSP-990
Solubility Information DMSO : ≥ 33 mg/mL (86.98 mM)
SMILES CC1=C2C(C[C@H](C3=C(C4=NC(OC)=CC=C4)C=C(F)C=C3)NC2=O)=NC(N)=N1
Molecular Weight (g/mol) 379.39
Formula Weight 379.39
Percent Purity 97.2%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.