missing translation for 'onlineSavingsMsg'
Learn More

MBP146-78, MedChemExpress

Product Code. 16413408
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
1mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16413408 10 mg 10mg
16499926 50 mg 50mg
16476127 5 mg 5mg
16470769 1 mg 1mg
16442756 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16413408 Supplier Medchem Express Supplier No. HY101525
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress MBP146-78 is a potent and selective inhibitor of cGMP dependent protein kinases.

MBP146-78 is a potent and selective inhibitor of cGMP dependent protein kinases.

Specifications

Chemical Name or Material MBP146-78
CAS 188343-77-3
Color White
Purity Grade Notes Research
Molecular Formula C21H22FN3
Quantity 10 mg
Solubility Information DMSO : 7.69 mg/mL (22.93 mM; Need ultrasonic)
SMILES CN1CCC(C2=CC(C3=CC=NC=C3)=C(C4=CC=C(F)C=C4)N2)CC1
Molecular Weight (g/mol) 335.42
Formula Weight 335.42
Percent Purity 99.91%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.