missing translation for 'onlineSavingsMsg'
Learn More

HO-3867, MedChemExpress

Product Code. 16423028
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16423028

Brand: Medchem Express HY100453

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress HO-3867 is a selective and potent STAT3 inhibitor and shows good antitumor activity.

HO-3867 is a selective and potent STAT3 inhibitor and shows good antitumor activity.

Specifications

Chemical Name or Material HO-3867
CAS 1172133-28-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C28H30F2N2O2
Quantity 10 mg
Solubility Information DMSO : ≥ 32 mg/mL (68.88 mM)
SMILES O=C1/C(CN(CC2=CC(C)(C)N(O)C2(C)C)C/C1=C\C3=CC=C(F)C=C3)=C/C4=CC=C(F)C=C4
Molecular Weight (g/mol) 464.55
Formula Weight 464.55
Percent Purity 98.26%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.