missing translation for 'onlineSavingsMsg'
Learn More

MAK683, MedChemExpress

Product Code. 16413888
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16413888

Brand: Medchem Express HY103663

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MAK683 is an embryonic ectoderm development (EED) inhibitor extracted from patent US20160176882 A1, compound example 2. MAK683 exhibits IC50s of 59, 89, 26 nM in EED Alphascreen binding, LC-MS and ELISA assay.

MAK683 is an embryonic ectoderm development (EED) inhibitor extracted from patent US20160176882 A1, compound example 2. MAK683 exhibits IC50s of 59, 89, 26 nM in EED Alphascreen binding, LC-MS and ELISA assay.

Specifications

Chemical Name or Material MAK683
CAS 1951408-58-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C20H17FN6O
Quantity 10 mg
Solubility Information DMSO : 52 mg/mL (138.15 mM; Need ultrasonic)
SMILES CC1=NC=CC=C1C2=CN=C(NCC3=C4CCOC4=CC=C3F)N5C2=NN=C5
Molecular Weight (g/mol) 376.39
Formula Weight 376.39
Percent Purity 99.27%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.