missing translation for 'onlineSavingsMsg'
Learn More

DMP 696, MedChemExpress

Product Code. 16414998
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16414998

Brand: Medchem Express HY12131

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress DMP 696 is a selective corticotropin-releasing hormone receptor 1 (CRHR1) antagonist, used for the treatment of anxiety and depression.

DMP 696 is a selective corticotropin-releasing hormone receptor 1 (CRHR1) antagonist, used for the treatment of anxiety and depression.

Specifications

Chemical Name or Material DMP 696
Physical Form Powder
CAS 202578-52-7
Color Light Brown
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C18H21Cl2N5O2
For Use With (Application) Metabolism-protein/nucleotide metabolism
Solubility Information DMSO : 100 mg/mL (243.72 mM; Need ultrasonic)
SMILES CC1=NN2C(N=C(C)N=C2NC(COC)COC)=C1C3=CC=C(Cl)C=C3Cl
Molecular Weight (g/mol) 410.3
Formula Weight 410.3
Percent Purity 98.73%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.