missing translation for 'onlineSavingsMsg'
Learn More

Disufenton sodium, MedChemExpress

Product Code. 16425588
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16425588

Brand: Medchem Express HY13244

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Disufenton sodium (NXY-059) is the disulfonyl derivative of the neuroprotective spin trap phenylbutynitrone(PBN), both NXY-059, its parent PBN and their hydrolysis/oxidation product MNT are very powerful scavengers of free radicals.

Disufenton sodium (NXY-059) is the disulfonyl derivative of the neuroprotective spin trap phenylbutynitrone(PBN), both NXY-059, its parent PBN and their hydrolysis/oxidation product MNT are very powerful scavengers of free radicals.

Specifications

Chemical Name or Material Disufenton sodium
CAS 168021-79-2
Color White
Purity Grade Notes Research
Molecular Formula C11H13NNa2O7S2
Quantity 10 mg
Synonym NXY-059
Solubility Information DMSO : 125 mg/mL (327.80 mM; Need ultrasonic) ∣H2O : ≥ 50 mg/mL (131.12 mM)
SMILES O=S(C1=C(C=CC(S(=O)(O[Na])=O)=C1)/C=[N+]([O-])/C(C)(C)C)(O[Na])=O
Molecular Weight (g/mol) 381.33
Formula Weight 381.33
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.