Learn More
MedChemExpress Diphenylterazine
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| CAS | 344940-63-2 |
| Solubility | DMSO : 27.5 mg/mL (72.86 mM∣Need ultrasonic∣DMSO can inactivate Diphenylterazine's activity) ∣DMF : 11.11 mg/mL (29.44 mM∣Need ultrasonic∣DMSO can inactivate Diphenylterazine's activity) ∣EtOH+HCl : 1 mg/mL (2.65 mM∣ultrasonic and warming and adjust pH to 2 with 1M HCl and heat to 80°C∣DMSO can inactivate Diphenylterazine's activity) ∣H2O : < 0.1 mg/mL (insoluble∣DMSO can inactivate Diphenylterazine's activity) |
| Synonym | DTZ |
| Content And Storage | -80°C, protect from light |
| Quantity | 10 mg |
| Molecular Weight (g/mol) | 377.44 g/mol |
| Product Type | Dye Reagents |
| Purity or Quality Grade | 95.0% |
| Shipping Condition | Room temperature |