Learn More
Tocris Bioscience™ CST 530
Click to view available options
:
10 mg
Description
CST 530 is a monovalent IAP antagonist. Induces cIAP1 and cIAP2 autodegradation in MM.1S cells (degrades cIAP1 at 0.1μM and cIAP2 at 1μM, after 16h). CST 530 reduces cell viability in a variety of cancer cell lines including SUDHL6, MOLM13 and HEL (IC50 values are 170 nM, 420 nM and 450 nM respectively).
Specifications
Specifications
| Chemical Name or Material | (2S,4S)-1-((S)-2-Cyclohexyl-2-((S)-2-(methylamino)propanamido)acetyl)-4-phenoxy-N-((R)-1,2,3,4-tetrahydronaphthalen-1-yl)pyrrolidine-2-carboxamide |
| CAS | 847137-53-5 |
| Quantity | 10 mg |
| Target | IAP Inhibitors |
| Molecular Formula | C33H44N4O4 |
| Purity | 97% |