missing translation for 'onlineSavingsMsg'
Learn More

10H-Phenoxazine, 98%, BLD Pharm™

Product Code. 31086880
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
10 g
25 g
100 g
500 g
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31086880 10 g
31086778 100 g
31076737 1 g
31076568 5 g
31084382 250 mg
31082048 500 g
31080793 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 31086880 Supplier BLD Pharm Supplier No. BD14749610g
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 135-67-1
Molecular Formula C12H9NO
MDL Number MFCD00005014
Quantity 10 g
SMILES C12=CC=CC=C1NC3=C(C=CC=C3)O2
Molecular Weight (g/mol) 183.21
Percent Purity 98%
Chemical Name or Material 10H-Phenoxazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.