Learn More
HLA DQ Antibody (rSPV-L3), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-08720AF532
Product Code. 18582854
  • 655.00 € / 100µL
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
HLA DQ Monoclonal antibody specifically detects HLA DQ in Human, Porcine samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Frozen)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HLA DQ
Monoclonal
Alexa Fluor 532
CD, CELIAC1DQ alpha 1 chain, DC-1 alpha chain, DQ-A1, FLJ27088, FLJ27328, GSE, HLA class II histocompatibility antigen, DQ(W3) alpha chain, HLA-DCA, HLA-DQA, leucocyte antigen DQA1, leukocyte antigen alpha chain, major histocompatibility complex, class II, DQ alpha 1, MGC149527, MHC class II antigen, MHC class II DQA1, MHC class II HLA-D alpha glycoprotein, MHC class II HLA-DQ-alpha-1, MHC class II surface glycoprotein, MHC HLA-DQ alpha
T4-positive CTL clone HG-38
100 μg
Adaptive Immunity, Asthma, Immunology
3117
Store at 4°C in the dark.
IgG1 κ
Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
rSPV-L3
50mM Sodium Borate
Mouse
Protein A or G purified
RUO
Primary
Human, Pig
Purified