Learn More
Human LAP (TGF-beta 1) Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody has been used in 8 publications
Brand:  R&D Systems MAB246-100
 View more versions of this product
Product Code. 18744259
  • 528.00 € / 100µg
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
LAP (TGF-beta 1) Monoclonal specifically detects LAP (TGF-beta 1) in Human samples. It is validated for Neutralization, Immunocytochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
LAP (TGF-beta 1)
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
TGFB1
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human LAP (TGF-beta 1)
100 μg
Primary
Detects human LAP in direct ELISAs and Western blots.
Human
Ascites
Neutralization, Immunocytochemistry
27235
Neutralization 1-5 ug/mL, Immunocytochemistry 3-25 ug/mL
CED, DPD1, LAP, LAP (TGFbeta 1), TGFB, TGFB1, TGFbeta, transforming growth factor beta 1
Mouse
Protein A or G purified from ascites
RUO
7040
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG1