missing translation for 'onlineSavingsMsg'
Learn More

rac 4-Hydroxy Ephedrine Hydrochloride, TRC

Product Code. 17963285
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17963285 100 mg 100mg
17993575 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17963285 Supplier Toronto Research Chemicals Supplier No. TRCH825305100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 942-51-8
Molecular Formula C10 H15 N O2 . Cl H
Molecular Weight (g/mol) 217.69
Synonym Oxilofrine hydrochloride, Benzenemethanol, 4-hydroxy-α-[1-(methylamino)ethyl]-, hydrochloride, (R*,S*)- (9CI), 4-Hydroxyephedrinehydrochloride, Suprifen hydrochloride, p-Hydroxyephedrine hydrochloride
IUPAC Name 4-[(1R,2S)-1-hydroxy-2-(methylamino)propyl]phenol;hydrochloride
SMILES Cl.CN[C@@H](C)[C@H](O)c1ccc(O)cc1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C10H15NO2.ClH/c1-7(11-2)10(13)8-3-5-9(12)6-4-8;/h3-7,10-13H,1-2H3;1H/t7-,10-;/m0./s1
Formula Weight 217.087
Percent Purity >95%
Chemical Name or Material Oxilofrine Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.