missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Birinapant, MedChemExpress
Description
Birinapant (TL32711) induces the autoubiquitylation and proteasomal degradation of cIAP1 and cIAP2 in intact cells, which results in formation of a RIPK1: caspase-8 complex, caspase-8 activation, and induction of tumor cell death. Birinapant (TL32711) targets TRAF2-associated cIAPs and abrogates TNF-induced NF-κB activation.
Specifications
Specifications
| Chemical Name or Material | Birinapant |
| CAS | 1260251-31-7 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C42H56F2N8O6 |
| Quantity | 100 mg |
| Synonym | TL32711 |
| Solubility Information | DMSO : 125 mg/mL (154.91 mM; Need ultrasonic) |
| SMILES | FC1=CC2=C(C=C1)C(C[C@H]3N(C([C@@H](NC([C@@H](NC)C)=O)CC)=O)C[C@@H](O)C3)=C(C(N4)=C(C[C@H]5N(C([C@@H](NC([C@@H](NC)C)=O)CC)=O)C[C@@H](O)C5)C6=C4C=C(F)C=C6)N2 |
| Molecular Weight (g/mol) | 806.94 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?