missing translation for 'onlineSavingsMsg'
Learn More

Autophinib, MedChemExpress

Product Code. 16424726
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16424726

Brand: Medchem Express HY101920

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Autophinib is a potent, selective autophagy inhibitor with IC50s of 90 nM and 40 nM for starvation- and Rapamycin-induced autophagy, respectively. Autophinib is also an ATP competitive Vacuolar Protein Sorting 34 (VPS34) inhibitor with an IC50 of 19 nM. Autophinib inhibits autophagy induced by starvation or Rapamycin by targeting VPS34.

Autophinib is a potent, selective autophagy inhibitor with IC50s of 90 nM and 40 nM for starvation- and Rapamycin-induced autophagy, respectively. Autophinib is also an ATP competitive Vacuolar Protein Sorting 34 (VPS34) inhibitor with an IC50 of 19 nM. Autophinib inhibits autophagy induced by starvation or Rapamycin by targeting VPS34.

Specifications

Chemical Name or Material Autophinib
CAS 1644443-47-9
Color Yellow
Purity Grade Notes Research
Molecular Formula C14H11ClN6O3
Quantity 100 mg
Solubility Information DMSO : ≥ 30 mg/mL (86.52 mM)
SMILES CC1=CC(NC2=NC(OC3=CC=C([N+]([O-])=O)C=C3)=NC(Cl)=C2)=NN1
Molecular Weight (g/mol) 346.73
Formula Weight 346.73
Percent Purity 99.56%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.