missing translation for 'onlineSavingsMsg'
Learn More

Antiblaze V6, TRC

Product Code. 17901205
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17901205 100 mg 100mg
17990915 25 mg 25mg
17943215 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17901205 Supplier Toronto Research Chemicals Supplier No. TRCA697400100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38051-10-4
Molecular Formula C13 H24 Cl6 O8 P2
Molecular Weight (g/mol) 582.99
Synonym Phosphoric acid, 2,2-bis(chloromethyl)-1,3-propanediyl tetrakis(2-chloroethyl) ester (9CI), Phosphoric acid, bis(2-chloroethyl) ester, diester with 2,2-bis(chloromethyl)-1,3-propanediol (7CI), 2,2-Bis(chloromethyl)-1,3-propanediol 2-chloroethanol phosphate (1:4:2), 2,2-Bis(chloromethyl)-1,3-propanediyl bis[bis(2-chloroethyl) phosphate], 2,2-Bis(chloromethyl)trimethylene bis[bis(2-chloroethyl) phosphate], 2,2-Bis-(chloromethyl)-propane-1,3-diyltetrakis(2-chloroethyl) bisphosphate, Amgard V 6, Antiblaze 100, Antiblaze AB 100, Antiblaze V 6, Celltech 60, 2,2-Bis(chloromethyl)-1,3-propanediol bis[di-(2-chloroethyl) phosphate], Phosgard 2XC20, Tetrakis(2-chlorethyl)dichloroisopentyldiphosphate, Tolgard V 6, V 6, V 6 (fireproofing agent), Phosphoric acid, 2,2-bis(chloromethyl)-1,3-propanediyl tetrakis(2-chloroethyl) ester (9CI), Phosphoric acid, bis(2-chloroethyl) ester, diester with 2,2-bis(chloromethyl)-1,3-propanediol (7CI), 2,2-Bis(chloromethyl)-1,3-propanediol 2-chloroethanol phosphate (1:4:2), 2,2-Bis(chloromethyl)-1,3-propanediyl bis[bis(2-chloroethyl) phosphate], 2,2-Bis(chloromethyl)trimethylene bis[bis(2-chloroethyl) phosphate], 2,2-Bis-(chloromethyl)-propane-1,3-diyltetrakis(2-chloroethyl) bisphosphate, Amgard V 6, Antiblaze 100, Antiblaze AB 100, Antiblaze V 6, Celltech 60, Phosgard 2XC20, Tetrakis(2-chlorethyl)dichloroisopentyldiphosphate, Tolgard V 6, V 6, V 6 (fireproofing agent)
IUPAC Name [2-[bis(2-chloroethoxy)phosphoryloxymethyl]-3-chloro-2-(chloromethyl)propyl] bis(2-chloroethyl) phosphate
SMILES ClCCOP(=O)(OCCCl)OCC(CCl)(CCl)COP(=O)(OCCCl)OCCCl

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C13H24Cl6O8P2/c14-1-5-22-28(20,23-6-2-15)26-11-13(9-18,10-19)12-27-29(21,24-7-3-16)25-8-4-17/h1-12H2
Formula Weight 579.9078
Percent Purity >95%
Chemical Name or Material Antiblaze V6
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.