missing translation for 'onlineSavingsMsg'
Learn More

4-[4-[(5S)-5-(Aminomethyl)-2-oxo-3-oxazolidinyl]phenyl]-3-morpholinone Hydrochloride, TRC

Product Code. 17901325
Change view
Click to view available options
Quantity:
10 mg
100 mg
1 g
Unit Size:
100mg
10mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17901325 100 mg 100mg
30134666 1 g 1g
17962295 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17901325 Supplier Toronto Research Chemicals Supplier No. TRCA616975100MG
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 898543-06-1
Molecular Formula C14 H17 N3 O4 . Cl H
Molecular Weight (g/mol) 327.76
Synonym 3-Morpholinone, 4-[4-[(5S)-5-(aminomethyl)-2-oxo-3-oxazolidinyl]phenyl]-, hydrochloride (1:1), 3-Morpholinone, 4-[4-[(5S)-5-(aminomethyl)-2-oxo-3-oxazolidinyl]phenyl]-, monohydrochloride (9CI), 4-[4-[(5S)-5-(Aminomethyl)-2-oxo-1,3-oxazolidin-3-yl]phenyl]morpholin-3-one hydrochloride
IUPAC Name 4-[4-[(5S)-5-(aminomethyl)-2-oxo-1,3-oxazolidin-3-yl]phenyl]morpholin-3-one;hydrochloride
SMILES Cl.NC[C@H]1CN(C(=O)O1)c2ccc(cc2)N3CCOCC3=O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C14H17N3O4.ClH/c15-7-12-8-17(14(19)21-12)11-3-1-10(2-4-11)16-5-6-20-9-13(16)18;/h1-4,12H,5-9,15H2;1H/t12-;/m0./s1
Formula Weight 327.0986
Chemical Name or Material 4-[4-[(5S)-5-(Aminomethyl)-2-oxooxazolidin-3-yl]phenyl]morpholin-3-one Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.