missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Manganese Bronze, CDA 863, C86300, 100g, ARMI∣MBH
Click to view available options
Quantity:
100 g
Unit Size:
100g
Description
Copper, Manganese Bronze, CDA 863, C86300, 100g
Specifications
Specifications
| Chemical Name or Material | Nickel/Silicon/Lead/Tin/Iron/Manganese/Aluminium/Zinc/Phosphorus/Carbon/Copper/Antimony/Arsenic/Cobalt |
| CAS | 7440-50-8 |
| Quantity | 100 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Nickel 0.02 %/Silicon 0.02 %/Lead 0.03 %/Tin 0.04 %/Iron 2.27 %/Manganese 3.72 %/Aluminium 6.03 %/Zinc 23.25 %/Phosphorus (0.004) %/Carbon (0.022) %/Copper (64.59) %/Antimony <0.01 %/Arsenic <0.01 %/Cobalt <0.01 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction