missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Silver Zirconium, NARLOY-Z, 100g, ARMI∣MBH
Click to view available options
Quantity:
100 g
Unit Size:
100g
Description
Copper, Silver Zirconium, NARLOY-Z, 100g
Specifications
Specifications
| Chemical Name or Material | Carbon/Zirconium/Silver/Copper/Aluminium/Chromium/Cobalt/Iron/Lead/Manganese/Nickel/Silicon/Tin/Zinc/Nitrogen/Oxygen/Phosphorus |
| CAS | 7440-50-8 |
| Quantity | 100 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Carbon 0.003 %/Zirconium 0.04 %/Silver 3.03 %/Copper 96.6 %/Aluminium (<0.01) %/Chromium (<0.01) %/Cobalt (<0.01) %/Iron (<0.01) %/Lead (<0.01) %/Manganese (<0.01) %/Nickel (<0.01) %/Silicon (<0.01) %/Tin (<0.01) %/Zinc (<0.01) %/Nitrogen [0.0002] %/Oxygen [0.0030] %/Phosphorus [0.004] % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction