missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cobalt, Co/Cr/Ni/Fe/Mo, Elgiloy, R30003, 100g, ARMI∣MBH
Click to view available options
Quantity:
100 g
Unit Size:
100g
Description
Cobalt, Co/Cr/Ni/Fe/Mo, Elgiloy, R30003, 100g
Specifications
Specifications
| Chemical Name or Material | Phosphorus/Boron/Nitrogen/Titanium/Copper/Niobium/Carbon/Tungsten/Oxygen/Vanadium/Tantalum/Aluminium/Silicon/Manganese/Molybdenum/Iron/Nickel/Chromium/Cobalt |
| CAS | 7440-48-4 |
| Quantity | 100 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Phosphorus 0.0019 %/Boron 0.002 %/Nitrogen 0.0034 %/Titanium 0.004 %/Copper 0.005 %/Niobium 0.006 %/Carbon 0.008 %/Tungsten 0.008 %/Oxygen 0.009 %/Vanadium 0.009 %/Tantalum 0.01 %/Aluminium 0.011 %/Silicon 0.05 %/Manganese 2.36 %/Molybdenum 8.6 %/Iron 12.6 %/Nickel 15.9 %/Chromium 20.4 %/Cobalt 40.4 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction