missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One 1000 μL Pipette Tips for Gilson™ Pipetman Pipettes
Description
- Manufactured from high-grade polypropylene
- Polypropylene has high breaking strength, dimensionally and stable heat-resistant
- Free of heavy metals
Specifications
Specifications
| Volume (Metric) | 200 to 1000 μL |
| Sterility | Non-sterile |
| Color | Blue |
| Autoclavable | Autoclavable |
| Certifications/Compliance | ISO 9001 |
| Format | Bulk |
| Tip Style | Extended Length |
| For Use With (Equipment) | Gilson Pipetman, Gilson Pipetman F250, Gilson Pipetman F300, Gilson Pipetman F500, Gilson Pipetman F1000, Gilson Pipetman U1000, Gilson Pipetman P1000 |
| Graduated | No |
| Material | Polypropylene |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?