Learn More
Human CD25/IL-2R alpha (Research Grade Basiliximab Biosimilar) Alexa Fluor 594-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Human Monoclonal Antibody
Brand:  R&D Systems FAB9926T-100UG
Product Code. 18721848
  • 507.00 € / 100µg
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
CD25/IL-2R alpha Monoclonal specifically detects CD25/IL-2R alpha in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD25/IL-2R alpha
Monoclonal
Alexa Fluor 594
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Human
Protein A or G purified from cell culture supernatant
Primary
Detects Human CD25/IL-2 R alpha based on Basiliximab therapeutic antibody. This non-therapeutic antibody uses the same variable region sequence as the therapeutic antibody Basiliximab. This product is for research use only.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
IgG1
Flow Cytometry
Hu107
Flow Cytometry 0.25-1 μg/106 cells
CD25, CD25 antigen, IDDM10, IL-2 R alpha, IL-2 receptor subunit alpha, IL2R, IL2R alpha, IL-2R subunit alpha, IL-2Ra, IL2RA, IL-2-RA, IL2-RA, interleukin 2 receptor, alpha, interleukin-2 receptor subunit alpha, p55, TAC antigen, TCGFR
Human CD25/IL-2 R alpha
100 μg
3559.0
Human
Supernatant