Learn More
Ibidi 10 mg/mL Rat Tail Collagen Type I
10 mg/mL Rat Tail Collagen Type I is a figh-quality rat tail collagen type I solution for cell culture applications.
387.20 € - 4646.40 €
Specifications
Product Type Rat Tail Collagen
Density 10 mg/mL
View More Specs
Products 3
Product Code Brand Volume (Metric) Quantity Price Quantity & Availability  
Product Code Brand Volume (Metric) Quantity Price Quantity & Availability  
17286123
View Documents Promotion Details
Ibidi
50204
5 mL 1
387.20 €
Each
View alternative products
Save Up to 
»
Estimated Shipment: 19-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
17296123
View Documents Promotion Details
Ibidi
50205
5 mL 4
1084.16 €
Each
View alternative products
Save Up to 
»
Estimated Shipment: 19-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
17206133
View Documents Promotion Details
Ibidi
50206
100 mL 1
4646.40 €
Each
View alternative products
Save Up to 
»
Estimated Shipment: 19-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description
Description
  • High-quality rat tail collagen type I solution for cell culture applications (e.g., scaffolds and bioprinting)
  • non-pepsinized
Specifications
Specifications
Rat Tail Collagen
10 mg/mL
Videos
SDS
Documents
Documents
Product Certifications