missing translation for 'onlineSavingsMsg'
Learn More

10-Deacetyl-7-xylosyl paclitaxel, MedChemExpress

Product Code. 16438018
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16438018 10 mg 10mg
16483937 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16438018 Supplier Medchem Express Supplier No. HY20584
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress 10-Deacetyl-7-xylosyl paclitaxel is a Paclitaxel (a microtubule stabilizing agent; enhances tubulin polymerization) derivative with improved pharmacological features.

10-Deacetyl-7-xylosyl paclitaxel is a Paclitaxel (a microtubule stabilizing agent

Specifications

Chemical Name or Material 10-Deacetyl-7-xylosyl paclitaxel
Physical Form Powder
CAS 90332-63-1
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C50H57NO17
For Use With (Application) Cancer-programmed cell death
Synonym 10-Deacetyl-7-xylosyltaxol 10-Deacetylpaclitaxel 7-Xyloside 10-Deacetyltaxol 7-Xyloside
Solubility Information DMSO : 33.33 mg/mL (35.31 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES CC(O[C@]([C@@]1([H])C[C@@H]2O[C@@](OC[C@@H](O)[C@@H]3O)([H])[C@@H]3O)(CO1)[C@]([C@@H]([C@]4(C(C)(C)C5=C(C)[C@@H](OC([C@H](O)[C@H](C6=CC=CC=C6)NC(C7=CC=CC=C7)=O)=O)C4)O)OC(C8=CC=CC=C8)=O)([H])[C@@]2(C([C@@H]5O)=O)C)=O
Molecular Weight (g/mol) 943.98
Formula Weight 943.98
Percent Purity 98.19%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.