Learn More
10,11-Dihydro-5-methyl-5H-dibenz[b,f]azepin-10-ol, TRC
Specifications
Specifications
| Chemical Name or Material | 10,11-Dihydro-5-methyl-5H-dibenz[b,f]azepin-10-ol |
| CAS | 28291-62-5 |
| Molecular Formula | C15H15NO |
| Quantity | 25 mg |
| InChI Formula | InChI=1S/C15H15NO/c1-16-13-8-4-2-6-11(13)10-15(17)12-7-3-5-9-14(12)16/h2-9,15,17H,10H2,1H3 |
| SMILES | OC1C=2C=CC=CC2N(C=3C=CC=CC3C1)C |
| IUPAC Name | 5-methyl-10,11-dihydro-5H-dibenzo[b,f]azepin-10-ol |
| Molecular Weight (g/mol) | 225.29 |
| Formula Weight | 225.12 |