Learn More
1-Trityl-3-methylpiperazine, TRC
Specifications
Specifications
| Chemical Name or Material | 1-Trityl-3-methylpiperazine |
| CAS | 326594-27-8 |
| Molecular Formula | C24H26N2 |
| Quantity | 1 g |
| Synonym | 3-Methyl-1-(triphenylmethyl)-piperazine |
| InChI Formula | InChI=1S/C24H26N2/c1-20-19-26(18-17-25-20)24(21-11-5-2-6-12-21,22-13-7-3-8-14-22)23-15-9-4-10-16-23/h2-16,20,25H,17-19H2,1H3 |
| SMILES | CC1CN(CCN1)C(c2ccccc2)(c3ccccc3)c4ccccc4 |
| IUPAC Name | 3-methyl-1-tritylpiperazine |
| Molecular Weight (g/mol) | 342.48 |
| Formula Weight | 342.2096 |