Learn More
1-Triacontylamine Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | 1-Triacontylamine Hydrochloride |
| CAS | 1346603-96-0 |
| Molecular Formula | C30H64ClN |
| Quantity | 500 mg |
| Synonym | 1-Triacontanamine Hydrochloride |
| InChI Formula | InChI=1S/C30H63N.ClH/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24-25-26-27-28-29-30-31;/h2-31H2,1H3;1H |
| SMILES | Cl.CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCN |
| IUPAC Name | triacontan-1-amine;hydrochloride |
| Molecular Weight (g/mol) | 474.29 |
| Formula Weight | 473.4727 |