missing translation for 'onlineSavingsMsg'
Learn More

1-Phenylpiperazine hydrochloride, 99%

Product Code. p-7023438
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Product Code. Quantity unitSize
11488836 25 g 25g
11498836 100 g 100g
2 options
This item is not returnable. View return policy

Product Code. 11488836

Brand: Thermo Scientific Alfa Aesar A16995.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2210-93-7 | C10H15ClN2 | 198.694 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2210-93-7
Molecular Formula C10H15ClN2
Molecular Weight (g/mol) 198.694
MDL Number MFCD00012758
InChI Key NQNZNONJZASOKL-UHFFFAOYSA-N
Synonym 1-phenylpiperazine hydrochloride, 1-phenylpiperazinium chloride, piperazine, 1-phenyl-, hydrochloride, phenylpiperazine hydrochloride, 1-phenyl piperazine hydrochloride, piperazine, 1-phenyl-, monohydrochloride, phenylpiperazine, chloride, 1-phenylpiperazine hcl, c10h14n2.hcl, 1-phenylpiperazinehydrochloride
PubChem CID 75164
IUPAC Name 1-phenylpiperazine;hydrochloride
SMILES C1CN(CCN1)C2=CC=CC=C2.Cl

Specifications

Melting Point 247°C to 250°C
Odor Odorless
Quantity 25 g
UN Number UN2811
Formula Weight 198.7
Percent Purity 99%
Chemical Name or Material 1-Phenylpiperazine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.