Learn More
1-(Phenylmethyl)-L-histidine Methyl Ester Monohydrochloride, TRC
Product Code. 30425082
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30425082 10 mg -
30409441 100 mg 100mg
30422109 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30425082 Supplier Toronto Research Chemicals Supplier No. TRCP99479310MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 274927-61-6
Molecular Formula C14H18ClN3O2
Molecular Weight (g/mol) 295.76
IUPAC Name methyl (2S)-2-amino-3-(1-benzylimidazol-4-yl)propanoate;hydrochloride
SMILES Cl.COC(=O)[C@@H](N)Cc1cn(Cc2ccccc2)cn1
Specifications
InChI Formula InChI=1S/C14H17N3O2.ClH/c1-19-14(18)13(15)7-12-9-17(10-16-12)8-11-5-3-2-4-6-11;/h2-6,9-10,13H,7-8,15H2,1H3;1H/t13-;/m0./s1
Quantity 10 mg
Formula Weight 295.1088
Shipping Condition Room Temperature
Chemical Name or Material 1-(Phenylmethyl)-L-histidine Methyl Ester Monohydrochloride