Learn More
1-Phenyl-5-(pyridin-2-yl)-2(1H)-pyridone-d5, TRC
Specifications
Specifications
| Chemical Name or Material | 1-Phenyl-5-(pyridin-2-yl)-2(1H)-pyridone-d5 |
| CAS | 2733718-04-0 |
| Molecular Formula | C16 D5 H7 N2 O |
| Quantity | 1 mg |
| Synonym | 1'-Phenyl[2,3'-bipyridin]-6'(1'H)-one-d5, 5-(2-Pyridyl)-1-phenyl-1,2-dihydropyridin-2-one-d5 |
| InChI Formula | InChI=1S/C16H12N2O/c19-16-10-9-13(15-8-4-5-11-17-15)12-18(16)14-6-2-1-3-7-14/h1-12H/i1D,2D,3D,6D,7D |
| SMILES | [2H]c1c([2H])c([2H])c(N2C=C(C=CC2=O)c3ccccn3)c([2H])c1[2H] |
| IUPAC Name | 1-(2,3,4,5,6-pentadeuteriophenyl)-5-pyridin-2-ylpyridin-2-one |
| Molecular Weight (g/mol) | 253.31 |
| Formula Weight | 253.126 |