missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-1-propyne, 99%, Thermo Scientific Chemicals

Catalog Number p-3758676
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
673-32-5
C9H8
116.16
MFCD00009272
GHUURDQYRGVEHX-UHFFFAOYSA-N
1-phenyl-1-propyne, prop-1-yn-1-ylbenzene, benzene, 1-propynyl, methylphenylacetylene, 2-propynylbenzene, 1-phenylpropyne, 1-phenylpropyne-1, unii-448376bfqc, prop-1-ynyl-benzene, 1-methyl-2-phenylacetylene
69601
prop-1-ynylbenzene
CC#CC1=CC=CC=C1
673-32-5
C9H8
116.16
MFCD00009272
GHUURDQYRGVEHX-UHFFFAOYSA-N
1-phenyl-1-propyne, prop-1-yn-1-ylbenzene, benzene, 1-propynyl, methylphenylacetylene, 2-propynylbenzene, 1-phenylpropyne, 1-phenylpropyne-1, unii-448376bfqc, prop-1-ynyl-benzene, 1-methyl-2-phenylacetylene
69601
prop-1-ynylbenzene
CC#CC1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Color Yellow
Density 0.9200g/mL
Boiling Point 185.0°C
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5C≡CCH3
Refractive Index 1.5630 to 1.5650
Quantity 5 g
Beilstein 05, 514
Specific Gravity 0.92
Formula Weight 116.16
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1-Phenyl-1-propyne
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'