Learn More
1-Phenoxy-2-propanol, TRC
Product Code. 30413189
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30413189 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30413189 Supplier Toronto Research Chemicals Supplier No. TRCP301700250G
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-Phenoxy-2-propanol
CAS 770-35-4
Molecular Formula C9 H12 O2
Quantity 250 g
Synonym 1-Phenoxy-2-propanol, (±)-1-Phenoxy-2-propanol, 2-Hydroxypropyl phenyl ether, 2-Phenoxy-1-methylethanol, NSC 24015, Phenoxyisopropanol, Propylene phenoxetol, rac-1-Phenoxy-2-propanol, β-Phenoxyisopropanol
InChI Formula InChI=1S/C9H12O2/c1-8(10)7-11-9-5-3-2-4-6-9/h2-6,8,10H,7H2,1H3
SMILES CC(O)COc1ccccc1
IUPAC Name 1-phenoxypropan-2-ol
Molecular Weight (g/mol) 152.19
Formula Weight 152.0837