Learn More
1-Phenanthrenecarboxylic Acid, TRC
Product Code. 30411461
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30411461 100 mg 100mg
30426494 10 mg 10mg
30419167 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30411461 Supplier Toronto Research Chemicals Supplier No. TRCP294795100MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 27875-89-4
Molecular Formula C15H10O2
Molecular Weight (g/mol) 222.24
Synonym NSC 139679
IUPAC Name phenanthrene-1-carboxylic acid
SMILES OC(=O)c1cccc2c1ccc3ccccc23
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C15H10O2/c16-15(17)14-7-3-6-12-11-5-2-1-4-10(11)8-9-13(12)14/h1-9H,(H,16,17)
Formula Weight 222.0681
Chemical Name or Material 1-Phenanthrenecarboxylic Acid