missing translation for 'onlineSavingsMsg'
Learn More

1-Pentadecyne, 98%

Product Code. p-7043755
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
This item is not returnable. View return policy

Product Code. 11397735

Brand: Thermo Scientific Alfa Aesar L02540.04

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 765-13-9 | C15H28 | 208.39 g/mol

1-Pentadecyne is used in pharmaceutical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Pentadecyne is used in pharmaceutical research.

Solubility
Not miscible in water.

Notes
Store under inert gas. Air sensitive. Avoid excessive heat and light. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 765-13-9
Molecular Formula C15H28
Molecular Weight (g/mol) 208.39
MDL Number MFCD00015081
InChI Key DONJGKADZJEXRJ-UHFFFAOYSA-N
Synonym 1-pentadecyne, 1-penatdecyne, pentadec-1-in, tridecylacetylene, 1-pentadecyne 1g, 1-pentadecyne,
PubChem CID 69825
IUPAC Name pentadec-1-yne
SMILES CCCCCCCCCCCCCC#C

Specifications

Melting Point 10°C
Density 0.793
Boiling Point 129°C to 131°C (10 mmHg)
Flash Point >90°C (194°F)
Refractive Index 1.442
Quantity 2 g
Beilstein 1751009
Solubility Information Not miscible in water.
Formula Weight 208.39
Percent Purity 98%
Chemical Name or Material 1-Pentadecyne
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.