Learn More
1-Methylimidazole-d6, TRC
Specifications
Specifications
| CAS | 285978-27-0 |
| Molecular Formula | C4 D6 N2 |
| Quantity | 50 mg |
| Synonym | 1H-Imidazole-2,4,5-d3, 1-(methyl-d3)- (9CI, ACI), 1-(Methyl-d3)-1H-imidazole-2,4,5-d3 (ACI), 1-(Methyl-d3)-1H-imidazole-2,4,5-d3, N-Methylimidazole-d6, 1-Methyl-1H-imidazole-D6, 1-Methylimidazole-d6 |
| InChI Formula | InChI=1S/C4H6N2/c1-6-3-2-5-4-6/h2-4H,1H3/i1D3,2D,3D,4D |
| SMILES | [2H]c1nc([2H])n(c1[2H])C([2H])([2H])[2H] |
| IUPAC Name | 2,4,5-trideuterio-1-(trideuteriomethyl)imidazole |
| Molecular Weight (g/mol) | 88.1408 |
| Formula Weight | 88.0908 |
| Chemical Name or Material | 1-Methylimidazole-d6 |