Learn More
1-Methylfluoranthene, TRC
Product Code. 30137749
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137749 2.5 mg 2.5mg
30008456 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30137749 Supplier Toronto Research Chemicals Supplier No. TRCM3053002.5MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 25889-60-5
Molecular Formula C17 H12
Molecular Weight (g/mol) 216.28
Synonym 1-Methyl-fluoranthene, 1-Methylfluoranthene
IUPAC Name 1-methylfluoranthene
SMILES Cc1ccc2cccc3c4ccccc4c1c23
Specifications
Quantity 2.5 mg
InChI Formula InChI=1S/C17H12/c1-11-9-10-12-5-4-8-15-13-6-2-3-7-14(13)16(11)17(12)15/h2-10H,1H3
Formula Weight 216.0939
Chemical Name or Material 1-Methylfluoranthene