Learn More
1-Methylcyclopentanol, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | 1-Methylcyclopentanol |
| CAS | 1462-03-9 |
| Molecular Formula | C6H12O |
| Quantity | 5 g |
| Synonym | 1-Hydroxy-1-methylcyclopentane, 1-Methyl-1-cyclopentanol, 1-Methyl-1-hydroxycyclopentane, 1-Methylcyclopentan-1-ol, 1-Methylcyclopentyl Alcohol, NSC 23230 |
| InChI Formula | InChI=1S/C6H12O/c1-6(7)4-2-3-5-6/h7H,2-5H2,1H3 |
| SMILES | CC1(O)CCCC1 |
| IUPAC Name | 1-methylcyclopentan-1-ol |
| Molecular Weight (g/mol) | 100.16 |
| Formula Weight | 100.0888 |