Learn More
(1-Methylcyclohexanyl)methyl-4-nitrophenyl Ether, TRC
Specifications
Specifications
| Chemical Name or Material | (1-Methylcyclohexanyl)methyl-4-nitrophenyl Ether |
| CAS | 85002-76-2 |
| Molecular Formula | C14 H19 N O3 |
| Quantity | 50 mg |
| Synonym | Benzene, 1-[(1-methylcyclohexyl)methoxy]-4-nitro-, (1-Methylcyclohexanyl)methyl-4-nitrophenyl ether, (1-Methylcyclohexyl)methyl-4-nitrophenyl ether, 1-[(1-Methylcyclohexyl)methoxy]-4-nitrobenzene |
| InChI Formula | InChI=1S/C14H19NO3/c1-14(9-3-2-4-10-14)11-18-13-7-5-12(6-8-13)15(16)17/h5-8H,2-4,9-11H2,1H3 |
| SMILES | CC1(COc2ccc(cc2)[N+](=O)[O-])CCCCC1 |
| IUPAC Name | 1-[(1-methylcyclohexyl)methoxy]-4-nitrobenzene |
| Molecular Weight (g/mol) | 249.31 |
| Formula Weight | 249.1365 |