missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-2-(tri-n-butylstannyl)imidazole, 90+%, Thermo Scientific Chemicals

Product Code. 15479225
Change view
Click to view available options
Quantity:
250 mg
1 g
Förpackningsstorlek:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Quantity unitSize
15479225 250 mg 250mg
15428117 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 15479225 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer H51523.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

Denna artikel kan inte returneras. Se returpolicy

1-Methyl-2-(tri-n-butylstannyl)imidazole is used in pharmaceuticals, drug candidates, ligands for transition metal catalysts and other molecular functional materials. It is an organostannic compound for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Methyl-2-(tri-n-butylstannyl)imidazole is used in pharmaceuticals, drug candidates, ligands for transition metal catalysts and other molecular functional materials. It is an organostannic compound for proteomics research.

Solubility
Sparingly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 105494-69-7
Molecular Formula C16H32N2Sn
Molecular Weight (g/mol) 371.156
MDL Number MFCD01319030
InChI Key KFWFYOPKNSYTMV-UHFFFAOYSA-N
Synonym 1-methyl-2-tributylstannyl-1h-imidazole, 1-methyl-2-tributylstannyl imidazole, 1-methyl-2-tri-n-butylstannyl imidazole, 1-methyl-1h-imidazol-2-yl tributylstannane, 1-methyl-2-tributylstannanyl-1h-imidazole, acmc-20aotl, tributyl-1-methylimidazol-2-yl stannane, n-methyl 2-tributylstannylimidazole, 2-tri-n-butylstannyl-n-methylimidazole
PubChem CID 2763252
IUPAC Name tributyl-(1-methylimidazol-2-yl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C1=NC=CN1C

Specifikationer

Quantity 250 mg
UN Number UN2788
Formula Weight 371.15
Percent Purity ≥90%
Chemical Name or Material 1-Methyl-2-(tri-n-butylstannyl)imidazole

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.